missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total HSP60 DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC1800-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464395

  • £720.60 / Pack of 5


Estimated Shipment
29-03-2019
Add to basket

Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
3329
5 Plates
Human
Videos