missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total Flt-3/Flk-2 DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC912-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454845

  • £401.31 / Pack of 2


Estimated Shipment date
from Supplier 06-04-2020
Add to basket

Description

Description

The Human Total Flt-3/Flk-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human Flt-3/Flk-2/CD135.
Specifications

Specifications

ELISA
Biotin
2322
2 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
Human
Videos
Documents