missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total ERK1 DuoSet IC ELISA
Human Total ERK1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Product Type | ELISA Development Kit for intracellular proteins |
Quantity | 1 Kit (for 5 Plates) |
Gene ID (Entrez) | 5595 |
Species | Human |
Description
The Human Total ERK1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human ERK1.Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
1 Kit (for 5 Plates) | |
Human |
ELISA Development Kit for intracellular proteins | |
5595 |