missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total ErbB3/Her3 DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
Brand: R&D Systems™ DYC234-5
Code : Z1
Additional Details : Weight : 0.01000kg
Description
- Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
- Development protocols are provided to guide further assay optimization
- Assay can be customized to your specific needs
- Economical alternative to complete kits
Specifications
ELISA | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
1 Kit |
Biotin | |
2065 | |
5 Plates | |
Human |