missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total ER alpha/NR3A1 DuoSet IC, R&D Systems™

Human Total ER alpha/NR3A1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC5715-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454725

  • £773.22 / Pack of 5


Estimated Shipment date
from Supplier 31-03-2020
Add to basket

Description

Description

The Human Total ER alpha/NR3A1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human ER alpha/NR3A1.
Specifications

Specifications

ELISA
Biotin
2099
5 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
Human
Videos
Documents