missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total Annexin A2 DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
POA
Specifications
Assay Range | 312.50 to 20000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gene ID (Entrez) | 302 |
Product Type | ELISA Development Kit for intracellular proteins |
Conjugate | Biotin |
Description
ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. This kit is designed for the simultaneous detection and enumeration of IFN-γ/IL-2. Complete ELISpot kits are ready-to-run and require no ass
The Human Total Annexin A2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human Annexin A2.Specifications
312.50 to 20000 pg/mL | |
302 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit for intracellular proteins | |
Human |