missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human TNF-beta Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human TNF-beta.

Brand:  Invitrogen™ BMS202CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15591517

  • £360.00 / 96 tests


Estimated Shipment date
from Supplier 25-02-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TNF-beta
HRP-Conjugate anti-human TNF-beta monoclonal antibody
Human TNF-beta Standard concentrate, 1ng/mL upon dilution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

The assay detects recombinant, unglycosylated lymphotoxin with the same sensitivity as the natural, glycosylated protein, shows good correlation with the standard cytotoxicity bioassay, and is specific for biologically active TNF-beta without TNF-alpha cross reactivity.
Specifications

Specifications

HRP
4.6pg/mL
TNF beta
96 Tests
Serum
ELISA
100 μL
Lymphotoxin-alpha, TNF superfamily member 1
Human
Videos
Documents

For Research Use Only