missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human TNF-beta Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human TNF-beta.

£360.00 - £2175.00

Specifications

For Use With (Application) ELISA
Sample Size 100 μL
Synonym Lymphotoxin-alpha, TNF superfamily member 1
Target TNF beta
Conjugate HRP
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TNF-beta
HRP-Conjugate anti-human TNF-beta monoclonal antibody
Human TNF-beta Standard concentrate, 1ng/mL upon dilution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15591517 View Documents Invitrogen™
BMS202CE
96 Tests £360.00
96 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 16-04-2020
Add to basket
 
15531527 View Documents Invitrogen™
BMS202TENCE
10 x 96 Tests £2175.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 16-04-2020
Add to basket
 
Description

Description

The assay detects recombinant, unglycosylated lymphotoxin with the same sensitivity as the natural, glycosylated protein, shows good correlation with the standard cytotoxicity bioassay, and is specific for biologically active TNF-beta without TNF-alpha cross reactivity.
Specifications

Specifications

ELISA
Lymphotoxin-alpha, TNF superfamily member 1
HRP
4.6pg/mL
100 μL
TNF beta
Serum

For Research Use Only