missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human TNF-alpha total Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human TNF-alpha.

Brand:  Invitrogen™ BMS2034CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15501537

  • £359.99 / 96 tests


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TNF-alpha
Biotin-Conjugate anti-human TNF-alpha polyclonal antibody
Streptavidin-HRP
Human TNF-alpha Standard lyophilized, 3000pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
Cachectin, Tumor necrosis factor alpha, DIF
ELISA
5.0pg/mL
50μL
TNF alpha
Videos

For Research Use Only