missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TNF alpha, Recombinant Protein, eBioscience™
Recombinant Protein
£58.00 - £201.00
Specifications
Accession Number | P01375 |
---|---|
Concentration | 0.1mg/mL |
Formulation | PBS with no preservative; pH 7.2 |
Gene ID (Entrez) | 7124 |
Molecular Weight (g/mol) | 19kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15597516
|
Affymetrix eBioscience
14-8329-62 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15207918
|
Affymetrix eBioscience
14-8329-63 |
10μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15245323
|
Affymetrix eBioscience
14-8329-81 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Tumor Necrosis Factor-Alpha (TNF alpha) is a protein secreted by lipopolysaccharide-stimulated macrophages, and causes tumor necrosis when injected into tumor-bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single ∽17kDa band following SDS-PAGE under non-reducing conditions. Description: Human TNF-alpha is a 17kDa factor produced by macrophages, monocytes, neutrophils, CD4^+ T cells and NK cells. A 26kDa form of TNF-alpha is expressed as a membrane bound molecule. TNF-alpha is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF-alpha have been observed. Applications Reported: Recombinant human TNF alpha is biologically active and can induce cytolysis of L929 mouse cell line in culture. Applications Tested: The ED50 of this protein, as measured by L929 cytotoxicity assay, is less than or equal to 80 pg/mL. This corresponds to a specific activity of greater than or equal to 1.3 x 10e7 Units/mg. Source: E. coli expressed amino acids Val 77-Leu 233 of human TNF alpha accession # HUMTNFAB, N-terminal His-tag. Bioactivity: The ED50 of this protein, as measured by L929 cytotoxicity assay, is less than or equal to 80 pg/mL. This corresponds to a specific activity of greater than or equal to 1.3 x 10e7 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >95% as determined by SDS-PAGE. Molecular Weight: 19 kDa. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2μm post-manufacturing filtered.Specifications
P01375 | |
PBS with no preservative; pH 7.2 | |
19kDa | |
7.2 | |
E. coli | |
RUO | |
Less than 0.01ng/μg cytokine as determined by the LAL assay. | |
TNF | |
Human | |
Hu | |
Recombinant | |
Recombinant |
0.1mg/mL | |
7124 | |
Human TNF alpha | |
SDS-PAGE | |
-80°C | |
E. coli expressed amino acids Val 77-Leu 233 of human TNF- alpha accession # HUMTNFAB, N-terminal His-tag | |
Human TNF alpha | |
The ED50 of this protein, as measured by L929 cytotoxicity assay, is less than or equal to 80pg/mL. This corresponds to a specific activity of greater than or equal to 1.3 x 107 Units/mg. | |
Unconjugated | |
E. coli | |
Liquid | |
Affinity chromatography |
For Research Use Only.