missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TNF-alpha Quantikine HS ELISA Kit, R&D Systems™

Brand:  R&D Systems™ HSTA00D

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13437405

  • £583.65 / 96 tests

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 0.191pg/mL
Specificity: Natural and recombinant human TNF-alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
7124
1 Plate
0.191pg/mL
Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos