missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human TNF alpha ELISA Kits

Tumor Necrosis Factor Alpha Recombinant protein STNFA from Thermo Scientific™ specifically detects Tumor Necrosis Factor Alpha in samples, and it is validated for ELISA and WB

Brand:  Thermo Scientific™ EH3TNFA5

Code : PN

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10561394

  • £1396.12 / 1 set


Estimated Shipment
09-03-2018
Add to basket

Description

Description

These human TNF alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of tumor necrosis factor alpha (TNF-alpha, TNFα) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched TNFα antibody pairs and TNFα standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human TNFα (human tumor necrosis factor alpha)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 15.6 to 1000pg/mL

  • Sensitivity: < 2pg/mL

  • Precision: Dependent upon sample type and level (see instructions)

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12, IL-12p40, IL-13, IL-15, IL-16, IL-17, IL-18, TNFβ, Eotaxin, RANTES, GROα, GROβ, MCP-1, MCP-2, MCP-3, MCP-4, VEGF, GCSF, GMCSF, MIP-1α, MIP-1β, TGFβ, IFNγ, IFNα; mouse TNFα; rat TNFα; and bovine TNFα. TNFα Receptor types 1 and 2 tested at 40mg/mL did not interfere in this assay. Recombinant pig TNFα cross-reacted at 100%

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

Specifications

Specifications

15.6 to 1000pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
30X Wash Buffer, 200mL
Absorbance at 450nm minus absorbance at 550nm
<2pg/mL
50µL
5 x 96 microplate wells
Natural and recombinant human TNFa (human tumor necrosis factor alpha)
Human TNF alpha ELISA Kit
Complete ELISA kit with coated 96-well plates
Dependent upon sample type and level (see instructions)
5 plate kit
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with human IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12, IL-12p40, IL-13, IL-15, IL-16, IL-17, IL-18, TNFß, Eotaxin, RANTES, GROa, GROß, MCP-1, MCP-2, MCP-3, MCP-4, VEGF, GCSF, GMCSF, MIP-1a, MIP
3.5 hr.
Videos
Documentation