eBioscience™ Human TIMP-1 Platinum ELISA Kit

For the quantitative detection of human tissue inhibitior of metallo-proteinase-1 (TIMP-1).

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: BMS2018TEN

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TIMP-1
Biotin-Conjugate anti-human TIMP-1 polyclonal antibody
Streptavidin-HRP
Human TIMP-1 Standard lyophilized, 5ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Product Code. 15561497

Quantity Price
1 £ 2085.02 / Pack of 10
Estimated Shipment date
from Supplier 05-05-2017
Add to basket


Description and Specification

Specification

Sample Type Cell culture supernatant, serum, plasma (EDTA, heparin)
Synonym Tissue inhibitor of metalloproteinases, EPA
Conjugate Biotin
Target TIMP-1
Sample Size 20μL (1:100 prediluted)
Quantity 10 x 96 tests
Species Human
For Use With (Application) ELISA
Sensitivity 10.0pg/mL

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.