missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TGF-beta 2 Quantikine ELISA Kit, R&D Systems™

Human TGF-beta 2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DB250

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442585

  • £539.52 / 96 tests

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human TGF-beta 2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TGF-beta 2 in cell culture supernates (125 ul), serum (125 ul), edta plasma (125 ul), heparin plasma (125 ul), citrate plasma (125 ul).
Specifications

Specifications

31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
7042
1 Plate
7pg/mL
Cell Culture Supernates (125μL), Serum (125μL), EDTA Plasma (125μL), Heparin Plasma (125μL), Citrate Plasma (125μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (125uL), Serum (125uL), EDTA Plasma (125uL), Heparin Plasma (125uL), Citrate Plasma (125uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
Videos