missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TGF-beta 2 Quantikine ELISA Kit, R&D Systems™
Human TGF-beta 2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 31.20 - 2000pg/mL |
---|---|
Assay Sensitivity | 7pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
Description
The Human TGF-beta 2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TGF-beta 2 in cell culture supernates (125 ul), serum (125 ul), edta plasma (125 ul), heparin plasma (125 ul), citrate plasma (125 ul).Specifications
31.20 - 2000pg/mL | |
7pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |