missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TGF-beta 1 DuoSet, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DY240

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13463345

  • £578.52 / Pack of 15


Estimated Shipment
29-10-2018
Add to basket

Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit
1 Kit
Biotin
7040
15 Plates
Human
Videos