missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human sVEGF R1/Flt-1 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid-phase ELISA designed to measure human soluble VEGF R1 in cell culture supernates, serum, plasma, saliva, and follicular fluid.

Brand:  R&D Systems™ SVR100B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452896

  • £3519.18 / Pack of 6


Estimated Shipment date
from Supplier 27-03-2020
Add to basket

Description

Description

It contains Sf 21-expressed recombinant human VEGF R1/Fc Chimera and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human VEGF R1 showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring soluble VEGF R1
Specifications

Specifications

1 Pack
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
2321
Cell Culture Supernates (100 μL), Serum (100 μL), EDTA Plasma (100 μL), Saliva (100 μL), Follicular Fluid (100 μL)
Serum
HRP
ELISA
6 Plates
Videos
Documents