missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sTNF-R (80 kd) Platinum ELISA Kit, for in vitro diagnostic use

Provides a simple, rapid and highly sensitive method for the determination of soluble TNF-R (80 kd) levels in body fluids or cell culture supernatants.

£360.00 - £2180.00

Specifications

For Use With (Application) ELISA
Sample Size 10 μL
Synonym Tumor necrosis factor receptor 2, TNFR2, CD120b
Target sTNF Receptor, TNF Receptor
Conjugate HRP
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sTNF-R (80 kDa)
HRP-Conjugate anti-human sTNF-R (80 kDa) monoclonal antibody
Human sTNF-R (80 kDa) Standard, 10ng/mL
Control, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15561667 View Documents Invitrogen™
BMS211CE
96 Tests £360.00
96 tests View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-04-2020
Add to basket
 
15581667 View Documents Invitrogen™
BMS211TENCE
10 x 96 Tests £2180.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-04-2020
Add to basket
 
Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

ELISA
Tumor necrosis factor receptor 2, TNFR2, CD120b
HRP
0.10ng/mL
10 μL
sTNF Receptor, TNF Receptor
Urine

For Research Use Only