missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sTNF-R (80 kd) Platinum ELISA Kit, for in vitro diagnostic use

Provides a simple, rapid and highly sensitive method for the determination of soluble TNF-R (80 kd) levels in body fluids or cell culture supernatants.

Brand:  Invitrogen™ BMS211TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581667

  • £2180.00 / Pack of 10


Estimated Shipment date
from Supplier 25-02-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sTNF-R (80 kDa)
HRP-Conjugate anti-human sTNF-R (80 kDa) monoclonal antibody
Human sTNF-R (80 kDa) Standard, 10ng/mL
Control, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
0.10ng/mL
sTNF Receptor, TNF Receptor
10 x 96 Tests
Urine
ELISA
10 μL
Tumor necrosis factor receptor 2, TNFR2, CD120b
Human
Videos
Documents

For Research Use Only