missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sTNF-R (60 kd) Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sTNF-R (60 kd).

Brand:  Invitrogen™ BMS203CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15521547

  • £359.99 / 96 tests


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal polyclonal antibody to human sTNF-R (60 kDa)
HRP-Conjugate anti-human sTNF-R (60 kDa) monoclonal antibody
Human sTNF-R (60 kDa) Standard, 10ng/mL upon dilution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

  • Provides a simple, rapid and highly sensitive method for the determination of soluble TNF-R (60 kd) levels in body fluids or cell culture supernatants
  • This assay will help to clarify the possible diagnostic and prognostic value of circulating sTNF-R (60 kd) in various neoplastic and inflammatory diseases
Specifications

Specifications

HRP
96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
Tumor necrosis factor receptor 1, TNF-R1, CD120a
ELISA
0.05ng/mL
10μL
sTNF Receptor, TNF Receptor
Videos

For Research Use Only