missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sPSGL-1 Platinum ELISA Kit

For the quantitative detection of human PSGL-1.

Brand:  Invitrogen™ BMS255TEN

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15531837

  • £2102.54 / Pack of 10


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human PSGL-1
Biotin-Conjugate anti-human PSGL-1 monoclonal antibody
Streptavidin-HRP
Human PSGL-1 Standard lyophilized, 100 U/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
10 x 96 tests
Cell culture supernatant, serum
Human
P-selectin glycoprotein ligand 1, CD162 antigen
ELISA
1.0 U/mL
100μL (1:20 prediluted)
sCD162 (sPSGL-1)
Videos

For Research Use Only