missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SPAG5 Partial ORF (AAH00322, 1082 a.a. - 1191 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010615-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein associated with the mitotic spindle apparatus. The encoded protein may be involved in the functional and dynamic regulation of mitotic spindles. [provided by RefSeq]
Sequence: AETETKVLQEALAGQLDSNCQPMATNWIQEKVWLSQEVDKLRVMFLEMKNEKEKLMIKFQSHRNILEENLRRSDKELEKLDDIVQHIYKTLLSIPEVVRGCKELQGLLEFSpecifications
AAH00322 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AETETKVLQEALAGQLDSNCQPMATNWIQEKVWLSQEVDKLRVMFLEMKNEKEKLMIKFQSHRNILEENLRRSDKELEKLDDIVQHIYKTLLSIPEVVRGCKELQGLLEF | |
RUO | |
SPAG5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10615 | |
SPAG5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEEPEST/MAP126/hMAP126 | |
SPAG5 | |
Recombinant | |
wheat germ expression system |