missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sP-selectin Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sP-selectin. P-selectin belongs to the selectin family of adhesion molecules.

Brand:  Invitrogen™ BMS219-4CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581707

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sP-selectin
HRP-Conjugate anti-human sP-selectin monoclonal antibody
Human sP-selectin Standard lyophilized, 80ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
0.20ng/mL
CD62P (P-Selectin), sCD62P (sP-Selectin)
96 Tests
Serum
ELISA
10 μL
Granule membrane protein 140, PADGEM, CD62P
Human
Videos
Documents

For Research Use Only