Learn More
R&D Systems™ Human sP-Selectin/CD62P ELISA Kit, R&D Systems™
a 1.25 hour solid phase ELISA that measures sP-Selectin in cell culture supernates, serum, and plasma.
Brand:  R&D Systems™ BBE6
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13462475
  • £529.12 / 96 tests
Estimated Shipment
07-03-2019
Add to basket
Description
Description
It contains recombinant human sP-Selectin and antibodies raised against the recombinant factor. It has been shown to accurately quantitate the recombinant sP-Selectin. Results obtained using natural human sP-Selectin showed linear curves that were parallel to the standard curves obtained using the recombinant kit standards. These results indicate that this kit can be used to determine relative mass values for natural human sP-Selectin
Specifications
Specifications
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Human
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
6403
1 Plate
0.5ng/mL
Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos