missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human sL-Selectin/CD62L ELISA Kit, R&D Systems™

Brand:  R&D Systems™ SBBE4B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13432666

  • £3519.18 / Pack of 6


Estimated Shipment date
from Supplier 31-03-2020
Add to basket

Description

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Sensitivity: 0.3ng/mL
Specificity: Natural and recombinant human sL-Selectin
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

1 Pack
Human
6402
6 Plates
Cell Culture Supernates (10 μL), Serum (10 μL), EDTA Plasma (10 μL), Heparin Plasma (10 μL), Citrate Plasma (10 μL)
Serum
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Solid Phase Sandwich ELISA
0.3ng/mL
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents