Learn More
Invitrogen™ IL-6 Receptor (Soluble) Human ELISA Kit
For the quantitative detection of human sIL-6R.
Brand: Invitrogen™ BMS214TEN
2046.76 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sIL-6R
HRP-Conjugate anti-human sIL-6R monoclonal antibody
Human sIL-6R Standard lyophilized, 5ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Description
The Human Interleukin-6 Receptor (Hu IL-6R) ELISA quantitates Hu IL-6R in human serum, plasma, amniotic fluid, urine or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-6R. Principle of the method The Human IL-6R solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL6R (interleukin-6 receptor subunit alpha) is a part of the receptor for interleukin 6. The IL-6 receptor complex is composed of two membrane glycoproteins: the low affinity receptor and the signaltransducing component. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer.Specifications
P08887 | |
0.01 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
CD126,IL-6R-1,IL-6RA,IL6QA,IL6RQ,gp80,IL6R | |
1.7% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
2 hr. 10 min. |
0.08-5.0 ng/mL | |
HRP | |
Amniotic Fluid, Plasma, Serum, Supernatant, Urine | |
ELISA | |
3570 | |
2.2% | |
Pre-coated 96 well plates, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Amniotic Fluid, 20 μL; Plasma, 10 μL; Serum, 10 μL; Supernatant, 20 μL; Urine,20 μL | |
CD126, Membrane glycoprotein 80 | |
1 hr. 20 min. |
For Research Use Only