missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sIL-2R Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sIL-2R.

Brand:  Invitrogen™ BMS212-2TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15521677

  • £2175.09 / Pack of 10


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sIL-2R
HRP-Conjugate anti-human sIL-2R monoclonal antibody
Human sIL-2R Standard lyophilized, 40ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
CD25 antigen, TAC, Interleukin-2 receptor alpha
ELISA
0.27ng/mL
50μL
IL-2 Receptor, sIL-2 Receptor
Videos

For Research Use Only