missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human sE-Selectin/CD62E Quantikine ELISA Kit, R&D Systems™
Human sE-Selectin/CD62E Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 0.13 - 8ng/mL |
---|---|
Assay Sensitivity | 0.027ng/mL |
Sample Type | Cell Culture Supernatant, Plasma (Citrate, Heparin), Serum |
Description
The Human sE-Selectin/CD62E Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human E-Selectin/CD62E in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul), citrate plasma (10 ul).Specifications
0.13 - 8ng/mL | |
0.027ng/mL | |
Cell Culture Supernatant, Plasma (Citrate, Heparin), Serum |