missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD93 (Soluble) Human Uncoated ELISA Kit

Specifically engineered for accurate and precise measurement of mouse IgG total in serum, plasma and cell culture supernatants.

Brand:  Invitrogen™ 88-50340-77

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15550867

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Human sCD93 Standard (lyophilized): 800ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent Z
Detection enzyme
Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

CD93 (Soluble) Human Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat total IgG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Human sCD93 ELISA kit contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA).
Specifications

Specifications

3.3ng/mL
Human
20 x 96 Tests
CD93
Cell Culture Supernatant, Plasma, Serum
ELISA
soluble C1q receptor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only