Learn More
Invitrogen™ CD93 (Soluble) Human Uncoated ELISA Kit with Plates
For quantitative measurement of human sCD93
Brand: Invitrogen™ 88-50340-22
Code : NEW
Additional Details : Weight : 0.47500kg
Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Human sCD93 Standard (lyophilized): 800ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent Z
Detection enzyme
Tetramethylbenzidine (TMB) Substrate Solution
Description
CD93 (Soluble) HumanUncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat total IgG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Human sCD93 ELISA kit contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA).Specifications
3.3ng/mL | |
Human | |
2 x 96 Tests | |
CD93 |
Cell Culture Supernatant, Plasma, Serum | |
ELISA | |
soluble C1q receptor |
For Research Use Only