missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sCD44var (v6) Platinum ELISA Kit

For the quantitative detection of human sCD44var (v6).

Brand:  Invitrogen™ BMS210

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15551657

  • £352.46 / 96 tests


Estimated Shipment date
from Supplier 01-06-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44var (v6)
HRP-Conjugate anti-human sCD44var (v6) monoclonal antibody
Human sCD44var (v6) Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

The sCD44var(v6) ELISA detects all circulating CD44 isoforms comprising the sCD44var(v6) sequences
Specifications

Specifications

HRP
96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
VFF-7 Phagocytic glycoprotein I, Hermes antigen
ELISA
0.05ng/mL
20μL (1:20 prediluted)
sCD44var (v6)
Videos

For Research Use Only