eBioscience™ Human sCD44std Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sCD44std.

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: BMS209/2CE

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44std
HRP-Conjugate anti-human sCD44std monoclonal antibody
Human sCD44std Standard, 8ng/mL
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Product Code. 15531617

Quantity Price
1 £ 331.26 / 96 tests
Estimated Shipment date
from Supplier 04-05-2017
Add to basket


Description and Specification

Specification

Sensitivity 0.02ng/mL
Species Human
Target CD44std, sCD44std
Synonym Phagocytic glycoprotein I, Hermes antigen, Epican
For Use With (Application) ELISA
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine
Quantity 96 tests
Sample Size 20μL (1:60 prediluted)

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.