missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sCD44std Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sCD44std.

Brand:  Invitrogen™ BMS209-2TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15551617

  • £2227.72 / Pack of 10


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44std
HRP-Conjugate anti-human sCD44std monoclonal antibody
Human sCD44std Standard, 8ng/mL
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine
Human
Phagocytic glycoprotein I, Hermes antigen, Epican
ELISA
0.02ng/mL
20μL (1:60 prediluted)
CD44std, sCD44std
Videos

For Research Use Only