missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sCD40L Platinum ELISA Kit, extra sensitive, for in vitro diagnostic use

For the quantitative detection of human sCD40L.

Brand:  Invitrogen™ BMS293TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15571897

  • £2175.09 / Pack of 10


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD40L
HRP-Conjugate anti-human sCD40L monoclonal antibody
Human sCD40L Standard lyophilized, 20ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
TRAP, HIGM 1, T cell antigen Gp39, CD154
ELISA
0.01ng/mL
20 to 50μL
sCD154 (sCD40 Ligand), sCD40
Videos

For Research Use Only