missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human sCD30 Platinum ELISA Kit, for in vitro diagnostic use
For the quantitative detection of human sCD30.
Brand: Invitrogen™ BMS240CE
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD30
HRP-Conjugate anti-human sCD30 monoclonal antibody
Human sCD30 Standard lyophilized, 200ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
HRP | |
96 tests | |
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin) | |
Human | |
Lymphoid activation antigen CD30, Ki-1 antigen |
ELISA | |
0.3ng/mL | |
25μL | |
CD30, sCD30 |
For Research Use Only