missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human sCD26 Platinum ELISA Kit, for in vitro diagnostic use
For the quantitative detection of human sCD26.
Brand: Invitrogen™ BMS235CE
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD26
Biotin-Conjugate anti-human sCD26 monoclonal antibody
Streptavidin-HRP
Human sCD26 Standard, 500ng/mL upon dilution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
7.3ng/mL | |
Cell Culture Supernatant, Serum | |
ELISA | |
20 μL | |
CD26, sCD26 |
Biotin | |
Human | |
96 Tests | |
Dipeptidyl peptidase IV, ADCP 2, ADABP, TP 103 |
For Research Use Only