Learn More
Invitrogen™ CD26 (Soluble) Human ELISA Kit
For the quantitative detection of human sCD26.
Brand: Invitrogen™ BMS235
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD26
Biotin-Conjugate anti-human sCD26 monoclonal antibody
Streptavidin-HRP
Human sCD26 Standard, 500ng/mL upon dilution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Human Soluble CD26 ELISA quantitates Hu sCD26 in human serum, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu sCD26. Principle of the method The Human sCD26 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti CD3-induced IL - 2 production. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of malignant transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.Specifications
P27487 | |
7.3 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
ADABP,ADCP2,CD26,DPPIV,TP103 | |
4.6% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 10 min. |
15.6-500 ng/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
1803 | |
9.1% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Serum, 20 μL; Supernatant, 20 μL | |
Dipeptidyl peptidase IV, ADCP 2, ADABP, TP 103 | |
1 hr. 20 min. |
For Research Use Only