Learn More
Invitrogen™ CD23 (Soluble) Human ELISA Kit
For the quantitative detection of human sCD23.
£437.00 - £2790.00
Specifications
Accession Number | P06734 |
---|---|
Assay Range | 12.5-400 U/mL |
Assay Sensitivity | 6.8 U/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD23
Biotin-Conjugate anti-human sCD23 monoclonal antibody
Streptavidin-HRP
Human sCD23 Standard lyophilized, 400 U/ml upon reconstitution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15511767
|
Invitrogen™
BMS227-2TEN |
10 x 96 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15591757
|
Invitrogen™
BMS227-2 |
96 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The Human Soluble CD23 (Hu sCD23) ELISA quantitates Hu sCD23 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu sCD23. Principle of the method The Human sCD23 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
P06734 | |
6.8 U/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
BLAST-2,CD23,CD23A,CLEC4J,FCE2,IGEBF | |
4% | |
HRP | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Immunoglobulin E receptor low affinity II | |
1 hr. 20 min. |
12.5-400 U/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
2208 | |
6.3% | |
Pre-coated 96 well plate, Adhesive Plate Covers, Sample Diluent, Sample Diluent, Biotinylated Detection Antibody, Biotinylated Detection Antibody, Wash Buffer, Wash Buffer, Chromogen, Controls, Controls | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
For Research Use Only