missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human sAPO-1/Fas Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sAPO-1/Fas.

Brand:  Invitrogen™ BMS245CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15511827

  • £359.99 / 96 tests


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sAPO-1/Fas
Biotin-Conjugate anti-human sAPO-1/Fas monoclonal antibody
Streptavidin-HRP
Human sAPO-1/Fas Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum
Human
CD95, Fas receptor, APT 1, ALPS 1 A, TNFRSF6
ELISA
13.2pg/mL
10μL
CD95 (APO-1/Fas), sCD95 (sAPO-1/Fas)
Videos

For Research Use Only