missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Resistin Quantikine ELISA Kit, R&D Systems™

Human Resistin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DRSN00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462725

  • £621.81 / 96 tests


Estimated Shipment date
from Supplier 06-04-2020
Add to basket

Description

Description

The Human Resistin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Resistin in cell culture supernates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).
Specifications

Specifications

1 Kit
Human
HRP
ELISA
Solid Phase Sandwich ELISA
0.055ng/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Serum
0.16 - 10ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
56729
1 Plate
Cell Culture Supernates (100uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents