missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Resistin Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid-phase ELISA designed to measure human Resistin in cell culture supernates, serum, and plasma.

Brand:  R&D Systems™ SRSN00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462866

  • £3037.79 / Pack of 6


Estimated Shipment
28-02-2019
Add to basket

Description

Description

It contains NS0-expressed recombinant human Resistin and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human Resistin showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring Resistin
Specifications

Specifications

0.16 - 10ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
56729
6 Plates
0.055ng/mL
Cell Culture Supernates (100μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (100uL), Serum (20uL), EDTA Plasma (20uL), Heparin Plasma (20uL)
Human
Videos