Learn More
Invitrogen™ Resistin Human ELISA Kit
For the quantitative detection of human Resistin. Resistin is a hormone secreted by adipose tissue.
£451.00 - £3070.00
Specifications
Accession Number | Q9HD89 |
---|---|
Assay Range | 31-2,000 pg/mL |
Assay Sensitivity | 3.1 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human Resistin
Biotin-Conjugate anti-human Resistin polyclonal antibody
Streptavidin-HRP
Human Resistin Standard lyophilized, 4000pg/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15541557
|
Invitrogen™
BMS2040 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 01-05-2024
Log in to see stock. |
|||||
15551557
|
Invitrogen™
BMS2040TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
The Human Resistin ELISA quantitates Hu Resistin in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Resistin. Principle of the method The Human Resistin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
Q9HD89 | |
3.1 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
ADSF,FIZZ31,RSTN,XCP1,RETN | |
5.1% | |
HRP | |
Plasma, 15 μL; Serum, 15 μL; Supernatant, 50 μL | |
Cysteine-rich secreted protein FIZZ3, ADSF, RETN1 | |
1 hr. 20 min. |
31-2,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
56729 | |
8.1% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 10 min. |
For Research Use Only