Learn More
Invitrogen™ RANTES Human ELISA Kit
Measure levels of regulated upon activation, normal T-cell expressed and secreted (RANTES) in human serum, plasma, tissue extracts, urine and culture supernatant samples.
£518.00 - £2060.00
Specifications
Accession Number | P13501 |
---|---|
Assay Sensitivity | 2 pg/mL |
Description | Human RANTES ELISA Kit |
Conjugate | Biotin |
Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10108643
|
Invitrogen™
EHRNTS |
96 Tests |
£518.00
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10763115
|
Invitrogen™
EHRNTS5 |
5 x 96 Tests |
£2060.00
1 set |
In Stock
Log in to see stock. |
|||||
Description
Assay RANTES (regulated upon activation, normal T-cell expressed and secreted) levels in human serum, plasma, tissue extracts, urine and culture supernatant samples with our Invitrgen™ Human RANTES ELISA Kits.
These human RANTES ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of RANTES (regulated upon activation, normal T-cell expressed and secreted) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.Target: Natural and recombinant human RANTES (regulated upon activation, normal T-cell expressed and secreted)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: Absorbance at 450nm minus absorbance at 550nm
Assay Range: 51.2 to 2000pg/mL
Sensitivity: < 2pg/mL
Precision: Intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3 hours
Sample Size: 50μL
Specificity: No significant cross-reaction with human Eotaxin, GROα, GROβ, MIP-1α, MIP-1β, MCP-1, MCP-2, MCP-3, MCP-4, G-CSF, GM-CSF, IFNα, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12, IL-13, IL-15, IL-16, IL-17, TNFα or TNFβ, or rat RANTES
Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.
Specifications
P13501 | |
Human RANTES ELISA Kit | |
Colorimetric ELISA (TMB substrate) with Plates | |
Colorimetric Microplate Reader | |
D17S136E,RANTES,SCYA5,SIS-delta,SISd,TCP228,eoCP | |
<10% | |
Absorbance at 450nm minus absorbance at 550nm | |
RUO | |
Human | |
3 hr. |
2 pg/mL | |
Biotin | |
ELISA Kit | |
6352 | |
<12% | |
HRP | |
Intra-assay CV <10%; inter-assay CV <10% | |
2°C to 8°C | |
1 hr. 20 min. |