missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Pro-MMP-13 Quantikine ELISA Kit, R&D Systems™

Human Pro-MMP-13 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DM1300

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442685

  • £610.05 / 96 tests

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human Pro-MMP-13 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-13 in cell culture supernates (50 ul).
Specifications

Specifications

78.00 - 5000pg/mL
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
4322
1 Plate
21.3pg/mL
Cell Culture Supernates (50μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (50uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents