missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human PlGF Quantikine ELISA Kit, R&D Systems™
Human PlGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 15.60 - 1000pg/mL |
---|---|
Assay Sensitivity | 7pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine |
Description
The Human PlGF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PLGF in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).Specifications
15.60 - 1000pg/mL | |
7pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine |