Learn More
R&D Systems™ Human PlGF Quantikine ELISA Kit, R&D Systems™
Human PlGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ SPG00
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13442856
  • £3660.30 / Pack of 6
Estimated Shipment date
from Supplier 28-10-2019
Description
Description
The Human PlGF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PLGF in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).
Specifications
Specifications
15.60 - 1000pg/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules
5228
6 Plates
7pg/mL
Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Citrate Plasma (100μL), Urine (100μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (100uL), Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL), Citrate Plasma (100uL), Urine (100uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates