missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PLA2G5 (aa 21-48) Control Fragment Recombinant Protein

Product Code. 30180551
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180551 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30180551 Supplier Invitrogen™ Supplier No. RP99794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84150 (PA5-84150. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of the secretory phospholipase A2 family. It is located in a tightly-linked cluster of secretory phospholipase A2 genes on chromosome 1. The encoded enzyme catalyzes the hydrolysis of membrane phospholipids to generate lysophospholipids and free fatty acids including arachidonic acid. It preferentially hydrolyzes linoleoyl-containing phosphatidylcholine substrates. Secretion of this enzyme is thought to induce inflammatory responses in neighboring cells. Alternatively spliced transcript variants have been found, but their full-length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P39877
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5322
Name Human PLA2G5 (aa 21-48) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Ca2+-dependent phospholipase A2; calcium-dependent phospholipase A2; DKFZp686C2294; FRFB; group V phospholipase A2; GV-PLA2; hVPLA(2); MGC46205; Phosphatidylcholine 2-acylhydrolase 5; phospholipase A2 group V; phospholipase A2 group V; calcium-dependent phospholipase A2; phospholipase A2, group 5; phospholipase A2, group V; PLA2; PLA2-10; Pla2g5; sPLA2
Common Name PLA2G5
Gene Symbol PLA2G5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GLLDLKSMIEKVTGKNALTNYGFYGCYC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.