Learn More
R&D Systems™ Human Phospho-VEGFR2/KDR DuoSet IC ELISA
Human Phospho-VEGFR2/KDR DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DYC1766-5
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13454375
  • £773.22 / Pack of 5
Estimated Shipment date
from Supplier 30-03-2020
Add to basket
Description
Description
The Human Phospho-VEGFR2/KDR DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human VEGFR2/KDR/Flk-1.
Specifications
Specifications
ELISA
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
Human
CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
1 Kit
3791
5 Plates
Store the unopened product at 2°C to 8°C. Do not use past expiration date.