missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-VEGF R2/KDR DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC1766E

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13464375

  • £1824.95 / Pack of 15


Estimated Shipment
05-04-2019
Add to basket

Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
3791
15 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos