Learn More
Human Phospho-VEGF R1/Flt-1 DuoSet IC, R&D Systems™
Human Phospho-VEGFR1/Flt-1 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  RD Systems (Bio-Techne) DYC4170-5
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13444655
  • £654.05 / Pack of 5
Estimated Shipment date
from Supplier 01-10-2019
Description
Description
The Human Phospho-VEGFR1/Flt-1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human VEGFR1/Flt-1.
Specifications
Specifications
ELISA
2321
5 Plates
Human
Biotin
ELISA Development Kit for phosphorylated intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.